Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOP2A Mouse anti-Human, Clone: OTI5E6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF802515
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.Specifications
TOP2A | |
Monoclonal | |
Unconjugated | |
TOP2A | |
TOP2, TP2A | |
Mouse | |
Affinity Chromatography | |
RUO | |
7153 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P11388 | |
TOP2A | |
Human recombit protein fragment corresponding to amino acids 1100-1531 of human TOP2A produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction