Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOP2A Mouse anti-Human, Mouse, PE/Cy5.5, Clone: TOP2A/1361, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254546PECY55

Catalog No. N254546PEC5


Add to cart

Description

Description

TOP2A Monoclonal antibody specifically detects TOP2A in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

TOP2A
TOP2A/1361
Flow Cytometry
Conjugated, Purified
TOP2A
Recombinant human Topoisomerase II alpha fragment (aa1352-1493) (exact sequence is proprietary)
Protein A or G purified
Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Phospho Specific, Stem Cell Markers
Primary
7153.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit