Learn More
Restek™ Topaz Splitless Single Taper (Gooseneck) Inlet Liner
SDP
Features revolutionary technology and inertness to deliver next level True Blue Performance
$147.00 - $286.00
Specifications
Quantity 5/Pk.
Liner Material Topaz
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
06-611-326
SDP
View Documents
Restek™
23331
Bruker/Varian GCs
Pack of 5 for $147.00
Add to cart
06-611-317
SDP
View Documents
Restek™
23321
Shimadzu GCs
Pack of 5 for $286.00
Add to cart
Description
Description
  • Deactivation: Unbelievably low breakdown for accurate and precise low-level GC analyses
  • Reproducibility: Unbeatable manufacturing controls and QC testing for superior reliability across compound classes
  • Productivity: Unparalleled cleanliness for maximized GC uptime and lab throughput
  • 100% Satisfaction: If a liner does not perform to expectations, it will be replaced or account will be credited
  • Patented
Specifications
Specifications
5/Pk.
Topaz