Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Topical Starch, Powder, USP, Spectrum™

Click to view available options
Quantity:
200 lbs.
Packaging:
Fiber Drum

S1995, 9005-25-8, (C6H10O5)n

Spectrum™ Topical Starch, Powder, USP is used to relieve itching, irritation, burning, or discomfort caused by hemorrhoids. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS 9005-25-8
CAS Max % 1
Molecular Formula C12H22O11
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
Ignition Residue 0.005
Iron (Fe) 10 μg/g
Grade USP
Loss on Drying 0.14
Oxidizing Substances 0.00018
Packaging Fiber Drum
pH 4.5 to 7.0
Quantity 200 lbs.
Show More Show Less