Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Topiramate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
97240-79-4
C12H21NO8S
339.36
KJADKKWYZYXHBB-XBWDGYHZSA-N
[(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-6-yl]methyl sulfamate
CC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1
97240-79-4
C12H21NO8S
339.36
KJADKKWYZYXHBB-XBWDGYHZSA-N
[(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-6-yl]methyl sulfamate
CC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1

T1696, 97240-79-4, C12H21NO8S

Spectrum™ Topiramate, USP is used alone or with other medications to prevent and control seizures like epilepsy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.002
Specific Rotation -28.6° to -35.0°
Water 0.005
Quantity 100 g
CAS Max % 1
Percent Purity 98 to 102%
Grade USP