Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TopoisomeraseIIIalpha Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP267219

Catalog No. NBP267219


Add to cart

Description

Description

TopoisomeraseIIIalpha Polyclonal antibody specifically detects TopoisomeraseIIIalpha in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

TopoisomeraseIIIalpha
Unconjugated
TBS (pH7.4), 1%BSA, 50% Glycerol with 0.05% Sodium Azide
DNA topoisomerase 3-alpha, TOP3, topoisomerase (DNA) III alpha
Rabbit
IgG
100μL
Primary
7156
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:200
Purified
TOP3A
Peptide
Protein A purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit