Learn More
Selleck Chemical LLC
Topotecan HCl 100mg 119413-54-6 NSC609699, Nogitecan HCl, SKFS 104864A
Manufacturer: Selleck Chemical LLC S1231100MG
Topotecan HCl (NSC609699, Nogitecan, SKFS 104864A) is a topoisomerase I inhibitor for MCF-7 Luc cells and DU-145 Luc cells with IC50 of 13 nM and 2 nM in cell-free assays, respectively. Topotecan HCl induces autophagy and apoptosis.