Learn More
Thermo Scientific™ TopVision Agarose
TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Manufacturer:  Thermo Scientific™ R0492
 View more versions of this product
Catalog No. FERR0492
Add to cart
Description
Description
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.
Specifications
Specifications
500 g