Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOR/mTOR Mouse anti-Human, Mouse, Biotin, Clone: 649, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243657B

Catalog No. NBP243657B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TOR/mTOR
649
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MTOR
Recombinant protein encompassing a sequence within the C-terminus region of human mTOR. The exact sequence is proprietary.
Protein G purified
Autophagy, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, Macroautophagy, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control
Primary
2475
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit