Learn More
TOR1AIP2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 247691AP
Catalog No. 50-173-4530
Add to cart
Description
Description
One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.
Specifications
Specifications
TOR1AIP2
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H496
LULL1, Lumenal domain like LAP1, RP11 12M5.5, TOR1AIP2, torsin A interacting protein 2
Rabbit
IgG
150 μL
-20°C
Primary
163590
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
TOR1AIP2
Liquid
TOR1AIP2
TOR1AIP2 Fusion Protein Ag20126
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human