Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOR3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540655
Description
Peptide sequence: FHFPHPKYVD LYKEQLMSQI RETQQLCHQT LFIFDEAEKL HPGLLEVLGP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%; Zebrafish: 83%.
The function remains unknown.
Specifications
TOR3A | |
Polyclonal | |
Unconjugated | |
TOR3A | |
Adir; ADIR2; ATP-dependant interferon response protein 1; ATP-dependant interferon responsive; ATP-dependent interferon-responsive protein; si:ch211-198n5.8; Tor3a; Torsin family 3 member A; torsin family 3, member A; torsin-3A | |
Rabbit | |
Affinity chromatography | |
RUO | |
64222 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H497 | |
TOR3A | |
Synthetic peptide directed towards the middle region of human TOR3A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction