Learn More
Perkin Elmer LLC Demountable Quartz Torch - Black Mark  DFS Item
Manufacturer:  Perkin Elmer LLC N8152379
The demountable quartz torch uses a removable and replaceable injector. It is recommended for HF applications (sapphire or Pt injectors) or when the user prefers to remove the injector for cleaning without dismounting the torch. It's included in the Geological Sample Kit (N8150040) and the Productivity Configuration's Sample Introduction Kit (N8150036).
Catalog No. NC1480934