Learn More
Selleck Chemical LLCSupplier Diversity Partner Torsemide 25mg 56211-40-6 AC-4464, JDL-464  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S169825MG
Torsemide (AC-4464, JDL-464) is a pyridyl sulfonylurea with a chemical structure between that of traditional loop diuretics and Cl- channel blockers, used to treat hypertension.
Catalog No. 50-656-1