Learn More
Tosoh Bioscience Tosoh Bioscience AIA™-1800 Systems
180 tests per hour with any combination AIA ST assays
Manufacturer:  Tosoh Bioscience 019836
 View more versions of this product
Catalog No. 22-279-320
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Standard Model: CPU, monitor, printer, power conditioner (ABC PC 1200), UPS (1500), and accessory kit
Total Laboratory Automation Model: CPU, monitor, printer, power conditioner (ABC PC 1200), UPS (1500), and accessory kit
Description
Description
  • Random access and continuous process
  • True auto dilution from 2 to 625
  • Fully automated daily maintenance and record keeping
Key Features
  • Up to 288 conductive pipette tips; up to 18 different barcoded
  • secondary reagents
  • Up to 17 barcoded sample racks with any combination 170; barcoded primary tubes and sample cups
  • Windows™ operator interface
  • Automatic reschedule, automatic rerun and automatic reflex testing
  • Sample level sensing, sample clot detectio
Specifications
Specifications
AIA 1800 Standard Model
36.61 in.
49.21 in.
180 Tests/Hr.
64.96 in.
661.38 lb.
180 Tests
93cm
125cm
300kg
165cm
Documents
Accessories