Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA™-1800 Systems
180 tests per hour with any combination AIA ST assays
Specifications
Throughput | 180 Tests/hr. |
---|---|
Quantity | 180 Tests |
Depth (English) | 36.61 in. |
Depth (Metric) | 93 cm |
Height (English) | 49.21 in. |
Includes: Standard Model: CPU, monitor, printer, power conditioner (ABC PC 1200), UPS (1500), and accessory kit
Total Laboratory Automation Model: CPU, monitor, printer, power conditioner (ABC PC 1200), UPS (1500), and accessory kit
Description
- Random access and continuous process
- True auto dilution from 2 to 625
- Fully automated daily maintenance and record keeping
- Up to 288 conductive pipette tips; up to 18 different barcoded
- secondary reagents
- Up to 17 barcoded sample racks with any combination 170; barcoded primary tubes and sample cups
- Windows™ operator interface
- Automatic reschedule, automatic rerun and automatic reflex testing
- Sample level sensing, sample clot detectio
Specifications
180 Tests/hr. | |
36.61 in. | |
49.21 in. | |
64.96 in. | |
661.38 lb. |
180 Tests | |
93 cm | |
125 cm | |
165 cm | |
300 kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title