Learn More
Tosoh Bioscience AIA™-360 System
Ideal for physician's office lab, work stations for the small hospitals, cardiac and specialty testing, as well as a back-up to larger analyzers
Supplier:  Tosoh Bioscience 0019945
Request Bulk Order
Catalog No. 22-279-322
Item Discontinued This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
Explore available promotions
Description
Description
  • 36 tests/hr.
  • Results in 18 minutes
  • Up to 25 patient samples and 25 test cups at one time
  • Barcoding feature
  • On-board camera for analyte identification
  • On-board printer
  • Uses 10-minute incubation and AIA ST reagents
  • Test menu: cardiac markers, tumor markers, thyroid and reproductive hormones, anemia assays and more
  • Fully automated, continuous random access
  • Touch-screen interface: saves up to 300 assay results
Specifications
Specifications
36 Tests/hr.
36 Tests
40 cm
52 cm
40 cm
50/60 Hz
61 lb.
Fluorescence enzyme immunoassay
16 in.
21 in.
16 in.
100/240 VAC 50/60 Hz
100/240 VAC
29 kg
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories