Learn More
Tosoh Bioscience AIA-PACK™ Intact PTH Assay Kit
Patented technology provides quantitative measurement of Intact Parathyroid Hormone (iPH) in human serum and EDTA plasma on specific AIA system analyzers
Manufacturer:  Tosoh Bioscience 025313
 View more versions of this product
Catalog No. 22-281-301
Add to cart
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
  • Kidney marker assay
  • Patented Unit Dose Test Cups for use with specific Tosoh Automated Immunoassay Systems
  • Two-site immunoenzymometric assay
  • Single, unitized cups require no pre-mixing, no on-board refrigeration, and no waste
  • Dry reagent format ensures 60 to 90 day calibration stabilty
  • Bar-coded for easy test identification and inventory management
  • Test results in about 20 minutes
Specifications
Specifications
Intact parathyroid hormone in Human Serum and EDTA Plasma on specific AIA system analyzer
12 x 1mL
Calibrator Set
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For in vitro diagnostic use only.