Tosoh™ Bioscience AIA-PACK™ Test Cups - AFP (Alpha Fetoprotein)

Test cup technology provides consistent and accurate assay results since all assays run the same way, every time

Manufacturer: Tosoh Bioscience LLC 020652

 View more versions of this product

Catalog No. 21-279-76

  Check Availability
Add to Cart

    Specifications

    Product Type AFP Calibration Verification/Linearity Test Set
    Detectable Analytes alpha-fetoprotein
    For Use With (Application) Ataxia telangiectasia, hereditary tyrosinemia, neonatal hepatitis, atresia of the biliary tract, tumour of other tissues
    For Use With (Equipment) AIA system
    Measuring Range Up to 0.4mg/L
    Quantity 2mL
    Sample Size 15μL
    Sample Type Serum, Urine
    Sensitivity 0.002mg/L
    Shelf Life 1 year

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit