Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - AFP (Alpha Fetoprotein)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Manufacturer:  Tosoh Bioscience 020552
 View more versions of this product
Catalog No. 21-279-64
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
These tiny, bar coded test cups contain premeasured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems.
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
1 Set contents: SDS, 2 × 4mL and CVM, 2 × 2mL.
Specifications
Specifications
alpha-fetoprotein
AIA system
AFP Sample Diluting Solution
4mL