Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - AFP (Alpha Fetoprotein)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Manufacturer: Tosoh Bioscience 025252
Description
These tiny, bar coded test cups contain premeasured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems.
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
1 Set contents: SDS, 2 × 4mL and CVM, 2 × 2mL.
Specifications
alpha-fetoprotein | |
AIA system | |
Up to 100ng/mL | |
100 Pack | |
Serum |
Ataxia telangiectasia, hereditary tyrosinemia, neonatal hepatitis, atresia of the biliary tract, tumour of other tissues | |
AIA Test Cup | |
alpha-fetoprotein Assay Test Cup | |
25μL | |
1ng/mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title