Tosoh Bioscience AIA-PACK™ Test Cups – B12 Vitamin

Test cup technology provides consistent and accurate assay results since all assays run the same way, every time

Manufacturer: Tosoh Bioscience LLC 020293

 View more versions of this product

Catalog No. 21-279-9

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Vitamin B12 Assay Test Cup
    For Use With (Application) Vitamin B12 deficiency
    For Use With (Equipment) AIA system
    Format AIA test cup
    Sample Size 100μL
    Sample Type Serum
    Sensitivity 84pg/mL
    Detectable Analytes Vitamin B12
    Measuring Range Up to 2000pg/mL
    Quantity Each

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit