Learn More
Tosoh Bioscience AIA-PACK™ Test Cups – B12 Vitamin
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier: Tosoh Bioscience 020706
This item is not returnable.
View return policy
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Vitamin B12 | |
Vitamin B12 Pretreatment Set | |
Vitamin B12 deficiency | |
Each |
Safety and Handling
ShelfLife : 1 Year