Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - BMG (Beta-2-Microglobulin)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Manufacturer:  Tosoh Bioscience 020359
 View more versions of this product
Catalog No. 21-279-34
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
Specifications
Specifications
beta-2-microglobulin
AIA system
BMG (beta-2-Microglobulin) Calibrator Set
15μL
0.002mg/L
Serum BMG determination
Up to 0.4mg/L
1mL
Serum,Urine
1 Year