Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tosoh Bioscience AIA-PACK™ Test Cups — Breast Cancer Marker 27.29

Specially designed for no preparation use with AIA analyzers

Manufacturer:  Tosoh Bioscience 025202

 View more versions of this product

Catalog No. 22-279-240


Add to cart

Description

Description

For determination of Breast Cancer Antigen 27.29 using Tosoh Automated Immunoassay Systems.

  • Contain all necessary components for CA 27.29 analysis
  • Single unit dose requires no preprocessing
  • Format: Dry reagent with 90 day calibration stability for most assays
  • Designed for 10 minute incubation protocol
  • Shelf life: one year from date of manufacture
  • Bar-coded for easy identification and inventory

Specifications

Specifications

Prostate specific Antigen
AIA 2000, AIA 900, AIA 600 II, AIA 360 system
Up to 50ng/mL
75μL
0.01ng/mL
Breast carcinoma-associated Antigen coded by the Human MUC-1 gene is identified
AIA test cup
100 Tests
Serum
27.29 Assay Test Cup
Safety and Handling

Safety and Handling

ShelfLife : 1 year

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit