Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tosoh Bioscience AIA-PACK™ Test Cups — Breast Cancer Marker 27.29

Specially designed for no preparation use with AIA analyzers

Manufacturer:  Tosoh Bioscience 020502

 View more versions of this product

Catalog No. 22-279-242


Add to cart

Description

Description

For determination of Breast Cancer Antigen 27.29 using Tosoh Automated Immunoassay Systems.

  • Contain all necessary components for CA 27.29 analysis
  • Single unit dose requires no preprocessing
  • Format: Dry reagent with 90 day calibration stability for most assays
  • Designed for 10 minute incubation protocol
  • Shelf life: one year from date of manufacture
  • Bar-coded for easy identification and inventory

Specifications

Specifications

Breast cancer marker
AIA 2000, AIA 900, AIA 600 II, AIA 360 system
4 x 100mL
27.29 Sample Diluting Solution
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit