Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK™ Test Cups — Breast Cancer Marker 27.29
Specially designed for no preparation use with AIA analyzers
$225.00 - $3056.00
Catalog Number | Mfr. No. | Detectable Analytes | Quantity | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Quantity | Type | Price | Quantity & Availability | |||
22-279-241
|
Tosoh Bioscience
020302 |
CA 27.29 | 12 mL | 27.29 Calibrator Set |
Each for $415.00
|
|
|||
22-279-240
|
Tosoh Bioscience
025202 |
Prostate specific Antigen | 100 Tests | 27.29 Assay Test Cup |
Each for $3,056.00
|
|
|||
22-279-242
|
Tosoh Bioscience
020502 |
Breast cancer marker | 4 x 100 mL | 27.29 Sample Diluting Solution |
Each for $225.00
|
|
|||
Description
For determination of Breast Cancer Antigen 27.29 using Tosoh Automated Immunoassay Systems.
- Contain all necessary components for CA 27.29 analysis
- Single unit dose requires no preprocessing
- Format: Dry reagent with 90 day calibration stability for most assays
- Designed for 10 minute incubation protocol
- Shelf life: one year from date of manufacture
- Bar-coded for easy identification and inventory
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title