TOSOH™ Bioscience AIA-PACK Test Cups — Breast Cancer Marker 27.29

Specially designed for no preparation use with AIA analyzers

Manufacturer: Tosoh Bioscience LLC 020302

 View more versions of this product

Catalog No. 22-279-241

  Check Availability
Add to Cart

    Specifications

    Product Type 27.29 Calibrator Set
    For Use With (Equipment) AIA 2000, AIA 900, AIA 600 II, AIA 360 system
    Format AIA test cup
    Measuring Range Up to 50ng/mL
    Quantity 12mL
    Detectable Analytes CA 27.29
    For Use With (Application) Breast carcinoma-associated Antigen coded by the Human MUC-1 gene is identified
    Sample Size 75μL
    Sample Type Serum
    Sensitivity 0.01ng/mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit