Tosoh Bioscience AIA-PACK™ Test Cups - CA 125 (Epithelial Ovarian Cancer)

Test cup technology provides consistent and accurate assay results since all assays run the same way, every time

Manufacturer: Tosoh Bioscience LLC 025288

 View more versions of this product

Catalog No. 21-280-018

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Product Type CA-125 Assay Test Cup
    Detectable Analytes CA 125 Determinant
    For Use With (Application) Gynecological neoplasms primarily endometrial and fallopian tube carcinomas and certain tumors of the pancreas, liver, colon, breast and lung
    For Use With (Equipment) AIA system
    Format AIA test cup
    Measuring Range Up to 1000U/mL
    Quantity Each
    Sample Size 100μL
    Sample Type Serum
    Sensitivity 2U/mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit