Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - CA 125 (Epithelial Ovarian Cancer)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier: Tosoh Bioscience 020388
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
CA 125 | |
2 U/mL | |
Gynecological neoplasms primarily endometrial and fallopian tube carcinomas and certain tumors of the pancreas, liver, colon, breast and lung | |
Up to 1000U/mL |
Serum | |
CA-125 Calibrator Set | |
Each | |
100 μL |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title