Learn More
Tosoh Bioscience AIA-PACK Test Cups - CA 125 (Epithelial Ovarian Cancer)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
$153.47 - $348.79
Catalog Number | Mfr. No. | Detectable Analytes | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Product Type | Price | Quantity & Availability | ||||
21-279-193
|
Tosoh Bioscience
020588 |
CA 125 | CA-125 Sample Diluting Solution |
Pack of 4 for $153.47
|
|
21-279-190
|
Tosoh Bioscience
020388 |
CA 125 | CA-125 Calibrator Set |
Pack of 12 for $348.79
|
|
21-280-018
|
Tosoh Bioscience
025288 |
CA 125 Determinant | CA-125 Assay Test Cup |
Pack of 100 for $2,502.75
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
AIA system | |
Each |