Tosoh Bioscience AIA-PACK™ Test Cups – CEA (Carcinoembryonic Antigen)

Test cup technology provides consistent and accurate assay results since all assays run the same way, every time

Manufacturer: Tosoh Bioscience LLC 020354

 View more versions of this product

Catalog No. 21-279-37

  Check Availability
Add to Cart

    Specifications

    Product Type CEA Calibrator Set
    For Use With (Application) Non-neoplastic and benign diseases such as Alcoholic cirrhosis, hepatitis, ulcerative colitis and Crohn's disease
    For Use With (Equipment) AIA system
    Format AIA test cup
    Measuring Range Up to 100ng/mL
    Quantity 4mL
    Sample Size 100μL
    Sample Type Serum
    Sensitivity 0.5ng/mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit