Learn More
Tosoh Bioscience AIA-PACK Test Cups - CORT (Cortisol)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
21-279-32
|
Tosoh Bioscience
020387 |
Cortisol Calibrator Set |
Each for $325.89
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-58
|
Tosoh Bioscience
020587 |
Cortisol Sample Diluting Solution |
Pack of 4 for $116.09
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
TOSOH AIA system | |
4 x 4mL |