Tosoh Bioscience AIA-PACK™ Test Cups - FOL (Folate)

Test cup technology provides consistent and accurate assay results since all assays run the same way, every time

Manufacturer: Tosoh Bioscience LLC 020392

 View more versions of this product

Catalog No. 21-279-29

  Check Availability
Add to Cart

    Specifications

    Product Type Folate Calibrator Set
    Quantity 1mL
    Sample Type Serum
    Shelf Life 1 year
    Detectable Analytes Folate
    For Use With (Application) Vitamin B12 deficiency
    For Use With (Equipment) AIA system
    Measuring Range Up to 20ng/mL
    Sample Size 100μL
    Sensitivity 0.8ng/mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit