Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - FOL (Folate)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier: Tosoh Bioscience 020292
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on TOSOH AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Folate assay | |
Serum | |
Folate Assay Test Cup | |
Each | |
100 μL |
AIA Test Cup | |
0.8 ng/mL | |
Vitamin B12 deficiency | |
Up to 20ng/mL |