Learn More
Tosoh Bioscience AIA-PACK Test Cups - IRI (Insulin)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
$344.00
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
21-279167
|
Tosoh Bioscience
020360 |
12 x 1mL | Insulin Calibrator Set |
Each for $344.00
|
|
21-279169
|
Tosoh Bioscience
020560 |
4 x 4mL | Insulin Sample Diluting Solution |
Pack of 4 for $123.65
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on TOSOH AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
TOSOH AIA system |