Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - PSA (Prostate Specific Antigen)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier: Tosoh Bioscience 025263
This item is not returnable.
View return policy
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on TOSOH AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Prostate specific Antigen | |
Serum | |
PSA Assay Test Cup | |
5 trays of 20 cups | |
20 μL |
AIA Test Cup | |
0.1 ng/mL | |
Prostate cancer | |
Up to 100ng/mL |