Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK Test Cups - PSA (Prostate Specific Antigen)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
21-279-36
|
Tosoh Bioscience
020363 |
4mL | PSA Calibrator Set |
Each for $124.71
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-62
|
Tosoh Bioscience
020563 |
4 x 4mL | PSA Sample Diluting Solution |
Pack of 4 for $154.43
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-74
|
Tosoh Bioscience
020663 |
Each | PSA Calibration Verification/Linearity Test Set |
Each for $122.58
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-280-004
|
Tosoh Bioscience
025263 |
5 trays of 20 cups | PSA Assay Test Cup |
Pack of 100 for $3,482.55
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on TOSOH AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
1Set contents: SDS, 2 x 4mL and CVM, 2 x 2mL.
Specifications
AIA system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title