Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK Test Cups TSH 3G (Thyroid Stimulating Hormone 3rd Generation)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Specifications
For Use With (Application) | Test cup technology provides consistent and accurate assay results since all assays run the same way, every time |
---|
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
21-279-21
|
Tosoh Bioscience
020294 |
5 trays of 20 cups | TSH 3G test cup |
Pack of 200 for $1,080.98
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-47
|
Tosoh Bioscience
020394 |
12 x 1mL | TSH 3G calibrator set |
Each for $335.48
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-85
|
Tosoh Bioscience
020594 |
4 x 4mL | TSH 3G sample diluting solution |
Pack of 4 for $120.45
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title