Learn More
Tosoh Bioscience AIA-PACK Test Cups - TSH (Thyroid Stimulating Hormone)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
21-279-20C
|
Tosoh Bioscience
020251100 |
N/A | TSH Assay; 20 cups x 5 trays |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on TOSOH AIA systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
1Set contents: SDS, 2 x 4mL and CVM 2 x 2mL.