Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK™ Unit Dose Test Cups
Specially prepared reagents for a variety of assays using Tosoh AIA systems
Manufacturer: Tosoh Bioscience 025287
Description
For determination of analytes using Tosoh Automated Immunoassay Systems.
- Contain all necessary components for analysis
- Single unit dose requires no preprocessing
- Format: Dry reagent with 90 day calibration stability for most assays
- Designed for 10 minute incubation protocol
- Shelf life: One year from date of manufacture
- Bar-coded for easy identification and inventory
Order Info
Packs of 100 (20 cups x 5 trays).
Specifications
Cortisol | |
AIA system | |
Up to 60μg/dL | |
100 Pack | |
Serum |
Measurement of Serum Cortisol and evaluate hypothalamic-pituitaryadrenal function as well as clinical diagnosis | |
AIA Test Cup | |
Cortisol Assay Test Cup | |
10μL | |
0.2μg/dL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title