Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tosoh Bioscience AIA-PACK™ Unit Dose Test Cups

Specially prepared reagents for a variety of assays using Tosoh AIA systems

Manufacturer:  Tosoh Bioscience 025284

 View more versions of this product

Catalog No. 21-280-023

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

For determination of analytes using Tosoh Automated Immunoassay Systems.

  • Contain all necessary components for analysis
  • Single unit dose requires no preprocessing
  • Format: Dry reagent with 90 day calibration stability for most assays
  • Designed for 10 minute incubation protocol
  • Shelf life: One year from date of manufacture
  • Bar-coded for easy identification and inventory

Order Info

Packs of 100 (20 cups x 5 trays).

Specifications

Specifications

C-Peptide
AIA system
Up to 30ng/mL serum, 300ng/mL urine
100 Pack
Serum,Urine
C-Peptide measurements are also used as an additional means of evaluating Glucose tolerance Tests
AIA Test Cup
C-Peptide Assay Test Cup
20μL
0.2ng/mL Serum, 2ng/mL Urine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit