TOSOH™ Bioscience AIA-PACK Unit Dose Test Cups

Specially prepared reagents for a variety of assays using Tosoh AIA systems

Manufacturer: Tosoh Bioscience LLC 025295

 View more versions of this product

Catalog No. 21-280-026

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Product Type IgE II Assay Test Cup
    Detectable Analytes IgE II
    Measuring Range 4000IU/mL
    For Use With (Application) Allergic rhinitis, extrinsic asthma, urticaria and atopic eczema
    For Use With (Equipment) AIA system
    Format AIA test cup
    Quantity 100 Pack
    Sample Size 10μL
    Sample Type Serum
    Sensitivity 3IU/mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit