Learn More
Tosoh Bioscience Accessories for G8 Automated Glycohemoglobin Analyzer
For use with G8 Automated Glycohemoglobin Analyzer.
Manufacturer:  Tosoh Bioscience 021560290SL
 View more versions of this product
Catalog No. 22-279-344
  Check Availability
Add to cart
Specifications
Description
For use with G8 Automated Glycohemoglobin Analyzer.
Description & Specifications
Specifications
19 in.
50/60Hz
114 lb.
51.5kg
112cm
LCD
4μL
100/240VAC 50/60Hz
48.2cm
Whole Blood
100/240VAC
44 in.
NGSP
G8 Automated Glycohemoglobin Analyzer
290 Sample Loader
For HbA1c diabetes measurement