Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience Hemoglobin A1c Control for G8 Automated HPLC Analyzer
System specific control for HbA1c analysis
Supplier: Tosoh Bioscience 220232
Description
For use as a control for Hemoglobin A1c analysis with G8 Automated HPLC Analyzer.
- Control solution
- Two levels
- 4 x 0.25mL/level
Specifications
2°C to 8°C | |
Hemoglobin A1c Control | |
Normal, Abnormal | |
7 Days |
G8 automated HPLC analyzer | |
Measurements are used in the clinical management of diabetes to assess the long-term efficacy of diabetic control | |
4 x 0.25 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title