Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience Reagents for G7 Analyzers
For use with G7 Automated HPLC Analyzer.
$301.40 - $6965.10
Catalog Number | Mfr. No. | Kit Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Size | Price | Quantity & Availability | |||||
22-007-004
|
Tosoh Bioscience
018767 |
4 x 4mL |
Each for $383.40
|
|
21-279-176
|
Tosoh Bioscience
020019 |
4 x 0.2mL |
Pack of 4 for $404.70
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-175
|
Tosoh Bioscience
997050 |
8 x 0.2mL |
Pack of 8 for $455.82
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
21-279-177
|
Tosoh Bioscience
997683 |
4/Kit |
Pack for $6,965.10
|
|
21-279-186
|
Tosoh Bioscience
018431US |
2000mL |
Each for $301.40
|
|
22-007-500
|
Tosoh Bioscience
021446 |
1500mL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
22-007-015
|
Tosoh Bioscience
019552 |
800mL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
22-007-016
|
Tosoh Bioscience
019553 |
800mL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
22-007-017
|
Tosoh Bioscience
019554/DEL/USE22279347 |
800mL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
For use with G7 Automated HPLC Analyzer.Specifications
G7 Automated HPLC Analyzer |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title