Learn More
Tosoh Bioscience ST AIA-PACK™ 25-OH Vitamin D Assay
Aids in the determination of Vitamin D sufficiency
Manufacturer:  Tosoh Bioscience 025234
 View more versions of this product
Catalog No. 21-280-150
Add to cart
Description
Description
For use on Tosoh AIA System Analyzers
  • Measures 100% of 25-OH Vitamin D3 and D2
  • No premixing
  • No pre-measuring
  • No on-board refrigeration
  • Dry reagent ensures 90-day calibration stability
  • Minimal waste
  • Cost effective
  • Barcoded for easy identification/inventory management
Specifications
Specifications
TOSOH AIA system analyzer
Aids in the determination of Vitamin D sufficiency
25-OH Vitamin D test cup reagent
3 Months
20 Cups x 5 trays
100 Pack