Learn More
Tosoh Bioscience ST AIA-PACK 25-OH Vitamin D Assay
Aids in the determination of Vitamin D sufficiency
Manufacturer: Tosoh Bioscience 025334
Description
For use on Tosoh AIA System Analyzers
- Measures 100% of 25-OH Vitamin D3 and D2
- No premixing
- No pre-measuring
- No on-board refrigeration
- Dry reagent ensures 90-day calibration stability
- Minimal waste
- Cost effective
- Barcoded for easy identification/inventory management
Specifications
TOSOH AIA system analyzer | |
Aids in the determination of Vitamin D sufficiency | |
12 x 1mL |
3 Months | |
25-OH Vitamin D Calibration set |