Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience ST AIA-PACK 25-OH Vitamin D Assay
Aids in the determination of Vitamin D sufficiency
Manufacturer: Tosoh Bioscience 025734
Description
For use on Tosoh AIA System Analyzers
- Measures 100% of 25-OH Vitamin D3 and D2
- No premixing
- No pre-measuring
- No on-board refrigeration
- Dry reagent ensures 90-day calibration stability
- Minimal waste
- Cost effective
- Barcoded for easy identification/inventory management
Specifications
TOSOH AIA system analyzer | |
Aids in the determination of Vitamin D sufficiency | |
25-OH Vitamin D Pretreatment set |
3 Months | |
PR-1: 1 x 32mL, PR-2: 6 x 5mL | |
6 Pack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title