Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tosoh Bioscience ST AIA-PACK™ 25-OH Vitamin D Assay

Aids in the determination of Vitamin D sufficiency

$164.12 - $2470.80

Specifications

For Use With (Application) Aids in the determination of Vitamin D sufficiency
For Use With (Equipment) TOSOH AIA system analyzer
Shelf Life 3 Months
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
21-280-152
View Documents
Tosoh Bioscience
025734
6 Pack 25-OH Vitamin D Pretreatment set
Each for $241.76
Add to cart
 
21-280-150
View Documents
Tosoh Bioscience
025234
100 Pack 25-OH Vitamin D test cup reagent
Pack of 100 for $2,470.80
Add to cart
 
21-280-151
View Documents
Tosoh Bioscience
025334
12 x 1mL 25-OH Vitamin D Calibration set
Each for $325.36
Add to cart
 
21-280-153
View Documents
Tosoh Bioscience
025434
4 x 1mL Control Set
Pack of 4 for $164.12
Add to cart
 
Description

Description

For use on Tosoh AIA System Analyzers

  • Measures 100% of 25-OH Vitamin D3 and D2
  • No premixing
  • No pre-measuring
  • No on-board refrigeration
  • Dry reagent ensures 90-day calibration stability
  • Minimal waste
  • Cost effective
  • Barcoded for easy identification/inventory management

Specifications

Specifications

Aids in the determination of Vitamin D sufficiency
TOSOH AIA system analyzer
3 Months
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit