Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Total Bilirubin Reagents

Determine quantity of total bilirubin in serum with these reagents.

Specifications

CE Marker Yes
Linearity 0 to 340µmol/L
State at Room Temperature Liquid
View More Specs

Products 1
Catalog Number Description Price Quantity & Availability  
Catalog Number Description Price Quantity & Availability  
1245250
Accessories View Documents
Total Bilirubin (Diazonium Salt)
Pack of 2 for $151.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Determine quantity of total bilirubin in serum in vitro with Thermo Scientific™ Total Bilirubin Reagents.

Bilirubin is produced from the catabolism of the heme molecule. Total bilirubin consists of both conjugated (direct) and unconjugated (indirect) fractions. Increased levels are associated with conditions causing obstruction of the bile duct, hepatitis, cirrhosis, hemolytic disorders, physiological jaundice of the newborn and several inherited enzyme deficiencies.
  • Suitable for manual use or on automated clinical chemistry analyzers
  • Applications for both sample blanking and non-sample blanking methods available on request for a wide range of automated analyzers
  • Offers at least 21 days working reagent stability and various kit sizes.

Total Bilirubin-Diazonium Salt (Product No. 1245-250):

  • Stability: Stable Liquid

Total Bilirubin-Acid Diazo (Product Nos. TR32321 and TR32326):

  • Stability: 21 Days (Working)

Specifications

Specifications

Yes
0 to 340µmol/L
Liquid
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit